missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GYPC Protein

A cDNA sequence encoding the GYPC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12071-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979268

  • 131.00 € / 2µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GYPC Protein
Research Use Only
Recombinant Protein
Baculovirus
ADPMWSTRSP NSTAWPLSLE PDPGMASAST TMHTTTIAEP DPGMSGWPDG RMETSTPTIM HHHHHH
Greater than 95.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GYPC protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers