missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GZMH Protein
A cDNA sequence encoding the GZMH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12072-HIS-EC-10ug
Additional Details : Weight : 0.01000kg
Specifications
2999 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEIIGGHEAK PHSRPYMAFV QFLQEKSRKR CGGILVRKDF VLTAAHCQGS SINVTLGAHN IKEQERTQQF IPVKRPIPHP AYNPKNFSND IMLLQLERKA KWTTAVRPLR LPSSKAQVKP GQLCSVAGWG YVSMSTLATT LQEVLLTVQK DCQCERLFHG NYSRATEICV GDPKKTQTGF KGDSGGPLVC KDVAQGILSY GNKKGTPPGV YIKVSHFLPW IKRTMKRL | |
Greater than 85.0% as determined by SDS-PAGE. |
GZMH Protein | |
Research Use Only | |
GZMH | |
Human | |
His | |
GZMH protein solution (0.25 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |