missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HAAO Protein

A cDNA sequence encoding the HAAO was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12098-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979358

  • 203.00 € / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HAAO Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMERRLG VRAWVKENRG SFQPPVCNKL MHQEQLKVMF IGGPNTRKDY HIEEGEEVFY QLEGDMVLRV LEQGKHRDVV IRQGEIFLLP ARVPHSPQRF ANTVGLVVER RRLETELDGL RYYVGDTMDV LFEKWFYCKD LGTQLAPIIQ EFFSSEQYRT GKPIPDQLLK EPPFPLSTRS IMEPMSLDAW LDSHHRELQA GTPLSLFGDT YETQVIAYGQ GSSEGLRQNV DVWLWQLEGS SVVTMGGRRL SLAPDDSLLV LAGTSYAWER TQGSVALSVT QDPACKKPLG
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
HAAO protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers