missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HADH Protein

A cDNA sequence encoding the HADH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12099-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929368

  • 120.00 € / 5µg
Estimated Shipment: 13-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3033
Greater than 95% as determined by SDS-PAGE.
Research Use Only
HADH
Human
His
The HADH solution contains 20mM Tris-HCl pH 8.0, 0.1M NaCl and 20% glycerol.
HADH Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSSSSTASAS AKKIIVKHVT VIGGGLMGAG IAQVAAATGH TVVLVDQTED ILAKSKKGIE ESLRKVAKKK FAENPKAGDE FVEKTLSTIA TSTDAASVVH STDLVVEAIV ENLKVKNELF KRLDKFAAEH TIFASNTSSL QITSIANATT RQDRFAGLHF FNPVPVMKLV EVIKTPMTSQ KTFESLVDFS KALGKHPVSC KDTPGFIVNR LLVPYLMEAI RLYERGDASK EDIDTAMKLG AGYPMGPFEL LDYVGLDTTK FIVDGWHEMD AENPLHQPSP SLNKLVAENK FGKKTGEGFY KYK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers