missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HbA1c Protein

Product Code. p-7149974
Click to view available options
Quantity:
10 mg
100 μg
500 μg
Unit Size:
100µg
10mg
500µg
This item is not returnable. View return policy

Product Code. 15909428

Brand: enQuireBio™ QP12117100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the HbA1c was constructed and used to recombinantly synthesize the protein.

Specifications

Name HbA1c Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species Human
Protein Tag Untagged
Buffer The HbA1c is supplied in a proprietary buffer formulation having pH 8.0. The HbA1c was dispensed on the basis of HbA1c not total hemoglobin.
Purity or Quality Grade Greater than 96.0%.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.