missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDDC3 Protein
A DNA sequence encoding the Homo sapiens (Human) HDDC3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7880-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001273380.1 | |
HDDC3 Protein | |
Research Use Only | |
HDDC3 | |
Recombinant Protein | |
E. coli | |
GSEAAQLLEA ADFAARKHRQ QRRKDPEGTP YINHPIGVAR ILTHEAGITD IVVLQAALLH DTVEDTDTTL DEVELHFGAQ VRRLVEEVTD DKTLPKLERK RLQVEQAPHS SPGAKLVKLA DKLYNLRDLN RCTPEVKIQ | |
Greater than 90% as determined by SDS-PAGE. |
374659 | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |