missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDGFL1 Protein
A cDNA sequence encoding the HDGFL1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12207-2ug
Additional Details : Weight : 0.01000kg
Specifications
154150 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSAYGMP MYKSGDLVFA KLKGYAHWPA RIEHMTQPNR YQVFFFGTHE TAFLSPKRLF PYKECKEKFG KPNKRRGFSA GLWEIENNPT VQASDCPLAS EKGSGDGPWP EPEAAEGDED KPTHAGGGGD ELGKPDDDKP TEEEKGPLKR SAGDPPEDAP KRPKEAAPDQ EEEAEAERAA EAERAAAAAA ATAVDEESPF LVAVENGSAP SEPGLVCEPP QPEEEELREE EVADEEASQE WHAEAPGGGD RDSL | |
Greater than 95.0% as determined by SDS-PAGE. |
HDGFL1 Protein | |
Research Use Only | |
HDGFL1 | |
Human | |
His | |
HDGFL1 protein solution (0.25 mg/ml) containing Phosphate buffered saline, (pH 7.4) and 10% glycerol. |