missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heterogeneous nuclear ribonucleoprotein 1

A DNA sequence encoding the Homo sapiens (Human) Heterogeneous nuclear ribonucleoprotein L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6176-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031350

  • 518.00 € / 100µg
Estimated Shipment: 28-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001005335.1
Heterogeneous nuclear ribonucleoprotein 1
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
3191
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HNRNPL
Recombinant Protein
E. coli
GENYDDPHKT PASPVVHIRG LIDGVVEADL VEALQEFGPI SYVVVMPKKR QALVEFEDVL GACNAVNYAA DNQIYIAGHP AFVNYSTSQK ISRPGDSDDS RSVNSVLLFT ILNPIYSITT DVLYTICNPC GPVQRIVIFR KNGVQAMVEF DSVQSAQRAK ASLNGADIYS GCCTLKIEYA KPTRLNVFKN DQDTWDYTNP NLSGQGDPGS NPNKRQRQPP LLGDHPAEYG GPHGGYHSHY HDEGYGP
Videos
SDS
Documents
Certificates
Special Offers

Special Offers