missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human HGF (NS0 Expressed) Protein
Description
Measured by its ability to induce IL-11 secretion by Saos-2 human osteosarcoma cells. Hjertner, O. et al. (1999) Blood 94:3883. The ED50 for this effect is equal to or less than 4.00 ng/mL.
Specifications
Specifications
| Accession Number | P14210.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose. |
| Gene ID (Entrez) | 3082 |
| Molecular Weight (g/mol) | M.W. (Observed): 30-40 kDa ( beta chain) & 54-66 kDa ( alpha chain), under reducing conditions., M.W. (Theoretical): 54 kDa ( alpha chain) & 26 kDa ( beta chain) |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived human HGF protein Gln32-Arg494 alpha chain & Val495-Ser728 beta chain |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.50 EU per 1μg of the protein by the LAL method. |
| Gene Alias | deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?