missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human HGF (NS0 Expressed) Protein

Product Code. p-200006985
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 18786215

Brand: R&D Systems™ 11352HG100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Wild type

Measured by its ability to induce IL-11 secretion by Saos-2 human osteosarcoma cells. Hjertner, O. et al. (1999) Blood 94:3883. The ED50 for this effect is equal to or less than 4.00 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14210.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose.
Gene ID (Entrez) 3082
Molecular Weight (g/mol) M.W. (Observed): 30-40 kDa ( beta chain) & 54-66 kDa ( alpha chain), under reducing conditions., M.W. (Theoretical): 54 kDa ( alpha chain) & 26 kDa ( beta chain)
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived human HGF protein Gln32-Arg494 alpha chain & Val495-Ser728 beta chain
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.50 EU per 1μg of the protein by the LAL method.
Gene Alias deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute 10μg size at 100μg/mL in PBS. Reconstitute all the other sizes at 200μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein HGF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.