missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human His6-USP4 Protein

Product Code. 18748657
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18748657 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18748657 Supplier R&D Systems™ Supplier No. E596050
  • ONLINE EXCLUSIVE PRICE
    492.12 €
    438.90 € / 50µg
    Save 53.22 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Reaction conditions will need to be optimized for each specific application. We recommend an initial recombinant human USP4 concentration of 10-100 nM when using Ubiquitin-AMC or Ubiquitin-Rh110 substrates (U-550, U-555)

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13107
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 7.5, 100 mM NaCl, 1 mM TCEP, 10% (v/v) Glycerol
Gene ID (Entrez) 7375
Molecular Weight (g/mol) M.W. (theoretical): 109 kDa
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human USP4 protein Contains an C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Alias Deubiquitinating enzyme 4, EC 3.1.2.15, EC 3.4.19.12, MGC149848, MGC149849, ubiquitin carboxyl-terminal esterase 4, ubiquitin carboxyl-terminal hydrolase 4, ubiquitin specific peptidase 4 (proto-oncogene), ubiquitin specific protease 4 (proto-oncogene), ubiquitin thioesterase 4, Ubiquitin thiolesterase 4, ubiquitin-specific processing protease 4, Ubiquitin-specific-processing protease 4, Ubiquitous nuclear protein homolog, UNP, Unph, UNPUnph
Common Name USP4
Gene Symbol USP4
Biological Activity Reaction conditions will need to be optimized for each specific application. We recommend an initial recombinant human USP4 concentration of 10-100 nM when using Ubiquitin-AMC or Ubiquitin-Rh110 substrates (U-550, U-555)
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 21 (baculovirus)-derived Contains an C-terminal 6-His tag
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein USP4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.