missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histamine H1 receptor Protein

A DNA sequence encoding the Homo sapiens (Human) Histamine H1 receptor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6181-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051380

  • 2285.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000852.1
Histamine H1 receptor Protein
Research Use Only
HRH1
Recombinant Protein
E. coli
AKIYKAVRQH CQHRELINRS LPSFSEIKLR PENPKGDAKK PGKESPWEVL KRKPKDAGGG SVLKSPSQTP KEMKSPVVFS QEDDREVDKL YCFPLDIVHM QAAAEGSSRD YVAVNRSHGQ LKTDEQGLNT HGASEISEDQ MLGDSQSFSR TDSDTTTETA PGKGKLRSGS NTGLDYIKFT WKRLRSHSRQ YVSGLHMNRE RKAAKQ
Greater than 90% as determined by SDS-PAGE.
3269
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers