missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone-binding protein RBBP7 Protein

Product Code. 16089560
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16089560 100 μg 100µg
16099560 10 μg 10µg
16039570 50 μg 50µg
16019570 200 μg 200µg
16029570 500 μg 500µg
16009570 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16089560 Supplier enQuireBio™ Supplier No. QP7585ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Histone-binding protein RBBP7, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001185648.1
Gene ID (Entrez) 5931
Name Histone-binding protein RBBP7 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol RBBP7
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MASKEMFEDT VEERVINEEY KIWKKNTPFL YDLVMTHALQ WPSLTVQWLP EVTKPEGKDY ALHWLVLGTH TSDEQNHLVV ARVHIPNDDA QFDASHCDSD KGEFGGFGSV TGKIECEIKI NHEGEVNRAR YMPQNPHIIA TKTPSSDVLV FDYTKHPAKP DPSGECNPDL RLRGHQKEGY GLSWNSNLSG HLLSASDDHT VCLWDINAGP KEGKIVDAKA IFTGHSAVVE DVAWHLLHES LFGSVADDQK LMIWDTRSNT TSKPSHLVDA HTAEVNCLSF NPYSEFILAT GSADKTVALW DLRNLKLKLH TFESHKDEIF QVHWSPHNET ILASSGTDRR LNVWDLSKIG EEQSAEDAED GPPELLFIHG GHTAKISDFS WNPNEPWVIC SVSEDNIMQI WQMAENIYND EESDVTTSEL EGQGS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.