missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone H2A.V Protein

A DNA sequence encoding the Homo sapiens (Human) Histone H2A.V, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7750-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050571

  • 505.00 € / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036544.1
Histone H2A.V Protein
Research Use Only
H2AFV
Recombinant Protein
E. coli
MAGGKAGKDS GKAKAKAVSR SQRAGLQFPV GRIHRHLKTR TTSHGRVGAT AAVYSAAILE YLTAEVLELA GNASKDLKVK RITPRHLQLA IRGDEELDSL IKATIAGGGV IPHIHKSLIG KKGQQKTA
Greater than 90% as determined by SDS-PAGE.
94239
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers