missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HK3 Protein
A cDNA sequence encoding the HK3 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 3101 |
---|---|
Name | HK3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | HK3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969898
|
enQuireBio™
QP12272-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949898
|
enQuireBio™
QP12272-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959898
|
enQuireBio™
QP12272-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
3101 | |
Research Use Only | |
HK3 | |
Human | |
His | |
The protein (1 mg/ml) contains 20mM Tris pH 8.0 and 10% glycerol. |
HK3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDSIGSSGLR QGEETLSCSE EGLPGPSDSSE LVQECLQQFKVTRAQLQQI QASLLGSMEQ ALRGQASPAP AVRMLPTYVG STPHGTEQGD FVVLELGATG ASLRVLWVTL TGIEGHRVEP RSQEFVIPQE VMLGAGQQLF DFAAHCLSEF LDAQPVNKQGLQLGFSFSFP CHQTGLDRST LISWTKGFRC SGVEGQDVVQ LLRDAIRRQG AYNIDVVAVV NDTVGTMMGC EPGVRPCEVG LVVDTGTNAC YMEEARHVAV LDEDRGRVCV SVEWGSFSDD GALGPVLTTF DHTLDHESLN PGAQRFEKMI GGLYLGELVR LVLAHLARCG VLFGGCTSPA LLSQGSILLE HVAEMEDPST GAARVHAILQ DLGLSPGASD VELVQHVCAA VCTRAAQLCA AALAAVLSCL QHSREQQTLQ VAVATGGRVC ERHPRFCSVL QGTVMLLAPE CDVSLIPSVDGGGRGVAMVT AVAARLAAHR RLLEETLAPF RLNHDQLAAV QAQMRKAMAK GLRGEASSLR MLPTFVRATP DGSERGDFLA LDLGGTNFRV LLVRVTTGVQ ITSEIYSIPE TVAQGSGQQL FDHIVDCIVD FQQKQGLSGQ SLPLGFTFSF PCRQLGLDQG ILLNWTKGFK ASDCEGQDVV SLLREAITRR QAVELNVVAI VNDTVGTMMS CGYEDPRCEI GLIVGTGTNA CYMEELRNVAGVPGDSGRMC INMEWGAFGD DGSLAMLSTR FDASVDQASI NPGKQRFEKM ISGMYLGEIV RHILLHLTSL GVLFRGQQIQ RLQTRDIFKT KFLSEIESDS LALRQVRAIL EDLGLPLTSDDALMVLEVCQ AVSQRAAQLC GAGVAAVVEK IRENRGLEEL AVSVGVDGTL YKLHPRFSSL VAATVRELAP RCVV | |
Greater than 95.0% as determined by SDS-PAGE. |