missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-E Protein
A DNA sequence encoding the Escherichia coli (strain K12) Aerobic glycerol-3-phosphate dehydrogenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7048-1mg
This item is not returnable.
View return policy
Specifications
NP_005507.3 | |
HLA-E Protein | |
Research Use Only | |
HLA-E | |
Recombinant Protein | |
E. coli | |
GSHSLKYFHT SVSRPGRGEP RFISVGYVDD TQFVRFDNDA ASPRMVPRAP WMEQEGSEYW DRETRSARDT AQIFRVNLRT LRGYYNQSEA GSHTLQWMHG CELGPDRRFL RGYEQFAYDG KDYLTLNEDL RSWTAVDTAA QISEQKSNDA SEAEHQRAYL EDTCVEWLHK YLEKGKETLL HLEPPKTHVT HHPISDHEAT LRCWALGFYP AEITLTWQQD GEGHTQDTEL VETRPAGDGT FQKWAAVVVP SGEEQRYTCH VQHEGLPEPV TLRWKPASQP TIPI | |
Greater than 90% as determined by SDS-PAGE. |
3133 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |