missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMBS Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | HMBS Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSHMSGNGN AAATAEENSP KMRVIRVGTR KSQLARIQTD SVVATLKASY PGLQFEIIAM STTGDKILDT ALSKIGEKSL FTKELEHALE KNEVDLVVHS LKDLPTVLPP GFTIGAICKR ENPHDAVVFH PKFVGKTLET LPEKSVVGTS SLRRAAQLQR KFPHLEFRSI RGNLNTRLRK LDEQQEFSAI ILATAGLQRM GWHNRVGQIL HPEECMYAVG QGALGVEVRA KDQDILDLVG VLHDPETLLR CIAERAFLRH LEGGCSVPVA VHTAMKDGQL YLTGGVWSLD GSDSIQETMQ ATIHVPAQHE DGPEDDPQLV GITARNIPRG PQLAAQNLGI SLANLLLSKG AKNILDVARQ LNDAH |
| Buffer | The HMBS solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction