missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGB2 Protein
A cDNA sequence encoding the HMGB2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5220.00 €
Specifications
Name | HMGB2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969928
|
enQuireBio™
QP12282-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959928
|
enQuireBio™
QP12282-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949928
|
enQuireBio™
QP12282-1MG |
1 mg |
5220.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HMGB2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HMGB2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 30% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH TGSMGKGDPN KPRGKMSSYA FFVQTCREEH KKKHPDSSVN FAEFSKKCSE RWKTMSAKEK SKFEDMAKSD KARYDREMKN YVPPKGDKKG KKKDPNAPKR PPSAFFLFCS EHRPKIKSEH PGLSIGDTAK KLGEMWSEQS AKDKQPYEQK AAKLKEKYEK DIAAYRAKGK SEAGKKGPGR PTGSKKKNEP EDEEEEEEEE DEDEEEEDED EE | |
Greater than 90% as determined by SDS-PAGE. |