missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HMGCR Protein

A DNA sequence encoding the Homo sapiens (Human) HMGCR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6169-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16051300

  • 505.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000850.1
HMGCR Protein
Research Use Only
HMGCR
Recombinant Protein
E. coli
MTRGPVVRLP RACDSAEVKA WLETSEGFAV IKEAFDSTSR FARLQKLHTS IAGRNLYIRF QSRSGDAMGM NMISKGTEKA LSKLHEYFPE MQILAVSGNY CTDKKPAAIN WIEGRGKSVV CEAVIPAKVV REVLKTTTEA MIEVNINKNL VGSAMAGSIG GYNAHAANIV TAIYIACGQD AAQNVGSSNC ITLMEASGPT NEDLYISCTM PSIEIGTVGG GTNLLPQQAC LQMLGVQGAC KDNPGENARQ LARIVCGTVM AGELSLMAAL AAGHLVKSHM IHNRSKINLQ DLQGACTKKT
Greater than 90% as determined by SDS-PAGE.
3156
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers