missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HNRNPL Protein
A DNA sequence encoding the Homo sapiens (Human) HNRNPL, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_001005335.1 |
---|---|
Gene ID (Entrez) | 3191 |
Name | HNRNPL Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037791
|
enQuireBio™
QP9182-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077791
|
enQuireBio™
QP9182-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047791
|
enQuireBio™
QP9182-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067791
|
enQuireBio™
QP9182-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087791
|
enQuireBio™
QP9182-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057791
|
enQuireBio™
QP9182-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001005335.1 | |
HNRNPL Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3191 | |
Research Use Only | |
HNRNPL | |
Recombinant Protein | |
Yeast | |
IRGLIDGVVE ADLVEALQEF GPISYVVVMP KKRQALVEFE DVLGACNAVN YAADNQIYIA GHPAFVNYST SQKISRPGDS DDSRSVNSVL LFTILNPIYS ITTDVLYTIC NPCGPVQRIV IFRKNGVQAM VEFDSVQSAQ RAKASLNGAD IYSGCCTLKI EYAKPTRLNV FKNDQDTWDY TNPNLSGQGD PGSNPNKRQR QPPLLGDHPA EYGGPHGGYH SHYHDEGYGP PPPHYEGRRM GPPVGGH | |
Greater than 90% as determined by SDS-PAGE. |