missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homeobox protein VENTX Protein
A DNA sequence encoding the Homo sapiens (Human) Homeobox protein VENTX, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7496-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055283.1 | |
Homeobox protein VENTX Protein | |
Research Use Only | |
VENTX | |
Recombinant Protein | |
E. coli | |
MRLSSSPPRG PQQLSSFGSV DWLSQSSCSG PTHTPRPADF SLGSLPGPGQ TSGAREPPQA VSIKEAAGSS NLPAPERTMA GLSKEPNTLR APRVRTAFTM EQVRTLEGVF QHHQYLSPLE RKRLAREMQL SEVQIKTWFQ NRRMKHKRQM QDPQLHSPFS GSLHAPPAFY STSSGLANGL QLLCPWAPLS GPQALMLPPG SFWGLCQVAQ EALASAGASC CGQPLASHPP TPGRPSLGPA LSTGPRGLCA MPQTGDAF | |
Greater than 80% as determined by SDS-PAGE. |
27287 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |