missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HRV-1A Protein

A DNA sequence encoding the Homo sapiens (Human) HRV-1A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9561-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048481

  • 615.00 € / 100µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HRV-1A Protein
Research Use Only
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
100 μg
Not determined.
Recombinant Protein
Yeast
NPVENYIDEV LNEVLVVPNI KESHHTTSNS APLLDAAETG HTSNVQPEDA IETRYVITSQ TRDEMSIESF LGRSGCVHIS RIKVDYTDYN GQDINFTKWK ITLQEMAQIR RKFELFTYVR FDSEITLVPC IAGRGDDIGH IVMQYMYVPP GAPIPSKRND FSWQSGTNMS IFWQHGQPFP RFSLPFLSIA SAYYMFYDGY DGDNTSSKYG SVVTNDMGTI CSRIVTEKQK HSVVITTHIY HKAKHTKAWC PRPPRAVPYT HSHVTNYMPE TGDVTTAIVR RNTITTA
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers