missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSP27 Protein
A cDNA sequence encoding the HSP27 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12327-1mg
Additional Details : Weight : 0.01000kg
Specifications
HSP27 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MTERRVPFSL LRGPSWDPFR DWYPHSRLFD QAFGLPRLPE EWSQWLGGSS WPGYVRPLPP AAIESPAVAA PAYSRALSRQ LSSGVSEIRH TADRWRVSLD VNHFAPDELT VKTKDGVVEI TGKHEERQDE HGYISRCFTR KYTLPPGVDP TQVSSSLSPE GTLTVEAPMP KLATQSNEIT IPVTFESRAQ LGGPEAAKSD ETAAK | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The HSP27 protein solution contains 20mM Hepes pH 7.5, 1mM DTT and 100mM KCl. |