missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HSPA9 Protein

A cDNA sequence encoding the HSPA9 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12330-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960079

  • 131.00 € / 10µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HSPA9 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MASEAIKGAV VGIDLGTTNS CVAVMEGKQA KVLENAEGAR TTPSVVAFTA DGERLVGMPA KRQAVTNPNN TFYATKRLIG RRYDDPEVQK DIKNVPFKIV RASNGDAWVE AHGKLYSPSQ IGAFVLMKMK ETAENYLGHT AKNAVITVPA YFNDSQRQAT KDAGQISGLN VLRVINEPTA AALAYGLDKS EDKVIAVYDL GGGTFDISIL EIQKGVFEVK STNGDTFLGG EDFDQALLRH IVKEFKRETG VDLTKDNMAL QRVREAAEKA KCELSSSVQT DINLPYLTMD SSGPKHLNMK LTRAQFEGIV TDLIRRTIAP CQKAMQDAEV SKSDIGEVIL VGGMTRMPKV QQTVQDLFGR APSKAVNPDE AVAIGAAIQG GVLAGDVTDV LLLDVTPLSL GIETLGGVFT KLINRNTTIP TKKSQVFSTA ADGQTQVEIK VCQGEREMAG DNKLLGQFTL IGIPPAPRGV PQIEVTFDID ANGIVHVSAK DKGTGREQQI VIQSSGGLSK DDIENMVKNA EKYAEEDRRK KERVEAVNMA EGIIHDTETK MEEFKDQLPA DECNKLKEEI SKMRELLARK DSETGENIRQ AASSLQQASL KLFEMAYKKM ASEREGSGSS GTGEQKEDQK EEKQ
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The HSPA9 protein solution contains 20mM Tris-HCl, pH 8.0, 10% glycerol and 0.5mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers