missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HTRA2 Protein

A cDNA sequence encoding the HTRA2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12360-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960169

  • 203.00 € / 50µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HTRA2 Protein
Research Use Only
Recombinant Protein
E. coli
MAVPSPPPAS PPSQYNFIAD VVEKTAPAVV YIEILDRHPF LGREVPISNG SGFVVAADGL IVTNAHVVAD RRRVRVRLLS GDTYEAVVTA VDPVADIATL RIQTKEPLPT LPLGRSADVR QGEFVVAMGS PFALQNTITS GIVSSAQRPA RDLGLPQTNV EYIQTDAAID FGNAGGPLVN LDGEVIGVNT MKVTAGISFA IPSDRLREFL HRGEKKNSSS GISGSQRRYI GVMMLTLSPS ILAELQLREP SFPDVQHGVL IHKVILGSPA HRAGLRPGDV ILAIGEQMVQ NAEDVYEAVR TQSQLAVQIR RGRETLTLYV TPEVTEGSHH HHHH
Greater than 95.0% as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The protein (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 50mM NaCl, 1mM DTT, and 20% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers