missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IF Protein
A cDNA sequence encoding the IF was constructed and used to recombinantly synthesize the protein.
131.00 € - 6380.00 €
Specifications
Name | IF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15990199
|
enQuireBio™
QP12371-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970199
|
enQuireBio™
QP12371-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980199
|
enQuireBio™
QP12371-1MG |
1 mg |
6380.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein solution (0.7 mg/ml) contains 20mM HEPES pH 8.0, 100mM NaCl, 20% Glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MAWFALYLLS LLWATAGTST QTQSSCSVPS AQEPLVNGIQ VLMENSVTSS AYPNPSILIA MNLAGAYNLK AQKLLTYQLM SSDNNDLTIG QLGLTIMALT SSCRDPGDKV SILQRQMENW APSSPNAEAS AFYGPSLAIL ALCQKNSEAT LPIAVRFAKT LLANSSPFNV DTGAMATLAL TCMYNKIPVG SEEGYRSLFG QVLKDIVEKI SMKIKDNGII GDIYSTGLAM QALSVTPEPS KKEWNCKKTT DMILNEIKQG KFHNPMSIAQ ILPSLKGKTY LDVPQVTCSP DHEVQPTLPS NPGPGPTSAS NITVIYTINN QLRGVELLFN ETINVSVKSG SVLLVVLEEA QRKNPMFKFE TTMTSWGLVV SSINNIAENV NHKTYWQFLS GVTPLNEGVA DYIPFNHEHI TANFTQY | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |