missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IFN-alpha 7/IFNA7 Protein

Product Code. 18767156
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18767156 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18767156 Supplier R&D Systems™ Supplier No. 11079IF050
  • ONLINE EXCLUSIVE PRICE
    570.89 €
    509.25 € / 50µg
    Save 61.64 €
    10% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Measured in anti-viral assays using HeLa human cervical epithelial carcinoma cells infected with encephalomyocarditis (EMC) virus. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. The ED50 for this effect is 1.50-30.0 pg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01567.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3444
Molecular Weight (g/mol) MolecularWeight-observed: 18-22 kDa, under reducing conditions., MolecularWeight-theroretical: 20 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human IFN-alpha 7/IFNA7 protein Cys24-Asp189
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IFNA7, IFNalpha J1, IFN-alpha J1, interferon alpha-7, interferon alpha-J1, interferon, alpha 7
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IFN-alpha J1/IFNA7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.