missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IFN-gamma Biotinylated Protein

Product Code. 18796015
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18796015 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18796015 Supplier R&D Systems™ Supplier No. BT285050
  • ONLINE EXCLUSIVE PRICE
    693.67 €
    618.45 € / 50µg
    Save 75.22 €
    10% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Biotinylated

Measured by its binding ability in a functional ELISA. When Recombinant Human IFN-gamma R1/CD119 (Catalog # 673-IR/CF ) is immobilized at 3μg/mL (100μL/well), Biotinylated Recombinant Human IFN-gamma (Catalog # BT285) binds with an ED50 of 3.00-27.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number CAA31639.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween 80.
Molecular Weight (g/mol) M.W. (Observed): 14-20 kDa, under reducing conditions., M.W. (Theoretical): 16.9 kDa
Quantity 50 μg
Source E. coli-derived human IFN-gamma protein Gln24-Gln166, with a N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 3 months, -20 to -70 °C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Biotin
Species Human
Reconstitution Reconstitute at 0.2 mg/mL in sterile, deionized water.
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.