missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IFNA7 Protein
A cDNA sequence encoding the IFNA7 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 3444 |
---|---|
Name | IFNA7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IFNA7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15910219
|
enQuireBio™
QP12375-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15990209
|
enQuireBio™
QP12375-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15900219
|
enQuireBio™
QP12375-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
3444 | |
Research Use Only | |
IFNA7 | |
Human | |
His | |
IFNA7 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |
IFNA7 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMCDLPQ THSLRNRRAL ILLAQMGRIS PFSCLKDRHE FRFPEEEFDG HQFQKTQAIS VLHEMIQQTF NLFSTEDSSA AWEQSLLEKF STELYQQLND LEACVIQEVG VEETPLMNED FILAVRKYFQ RITLYLMEKK YSPCAWEVVR AEIMRSFSFS TNLKKGLRRK D | |
Greater than 90% as determined by SDS-PAGE. |