missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IGFBP-3 Protein

Product Code. 18718507
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18718507

Brand: R&D Systems™ 8874B3025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IGFBP-3 Protein is derived from Sf 9 (baculovirus). The Recombinant Human IGFBP-3 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P17936
For Use With (Application) Bioactivity
Gene ID (Entrez) 3486
Molecular Weight (g/mol) Observed M.W.: 34-41kDa, reducing conditions, Theoretical M.W.: 29kDa
Quantity 25 μg
Source Spodoptera frugiperda,Sf 9 (baculovirus)-derived human IGFBP-3 protein Gly28-Lys291 Accession &Num; P17936
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias acid stable subunit of the 140 K IGF complex, binding protein 29, binding protein 53, growth hormone-dependent binding protein, IBP-3, IBP3BP-53, IGF-binding protein 3, IGFBP3, IGFBP-3, insulin-like growth factor binding protein 3, insulin-like growth factor-binding protein 3
Common Name IGFBP-3
Gene Symbol IGFBP3
Biological Activity Measured by its ability to inhibit the biological activity of IGF-I or IGF-II on MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is typically 0.03-0.15μg/mL in the presence of 14ng/mL Recombinant Human IGF-II.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 9 (baculovirus)-derived Gly28-Lys291 Accession # P17936
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.