missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-12 Protein
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
25µg
5µg
Description
Measured in a cell proliferation assay using PHA-stimulated human T lymphoblasts. Symons, J.A. et al. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 272. The ED50 for this effect is 0.01-0.05 ng/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 3592 |
| Molecular Weight (g/mol) | 34.7 kDa (p40) & 22.5 kDa (p35) |
| Name | IL-12 |
| Quantity | 5 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-12 protein, (N-terminus) Human IL-12 p40 (Ile23-Ser328) Accession &Num; P29460 Human IL-12 p35 (Arg23-Ser219) Accession &Num; P29459 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto