missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-15 Biotinylated Protein

Product Code. 18756305
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18756305 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18756305 Supplier R&D Systems™ Supplier No. BTN015025
  • ONLINE EXCLUSIVE PRICE
    1058.92 €
    942.90 € / 25µg
    Save 116.02 €
    10% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Biotinylated

Measured by its binding ability in a functional ELISA. When Recombinant Human IL-15R alpha Fc Chimera (HEK293) (Catalog # 7194-IR ) is immobilized at 0.25 μg/mL (100 μL/well), Biotinylated Recombinant Human IL-15 (Catalog # BTN015) binds with an ED50 of 0.0800-0.640 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40933.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3600
Molecular Weight (g/mol) MolecularWeight-observed: 8-14 kDa, under reducing conditions., MolecularWeight-theroretical: 13 kDa
Quantity 25 μg
Source E. coli-derived human IL-15 protein Asn49-Ser162
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IL15, IL-15 mgC9721, interleukin 15, interleukin-15
Conjugate Biotin
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.