missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-15 Protein, CF
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
Unit Size:
100µg
1mg
250µg
25µg
Description
Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. The ED50 for this effect is 0.300-2.60 ng/mL.
Specifications
Specifications
| Accession Number | P40933.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3600 |
| Molecular Weight (g/mol) | M.W. (Observed): 9 kDa, under reducing conditions., M.W. (Theoretical): 13 kDa |
| Quantity | 250 μg |
| Source | E. coli-derived human IL-15 protein Asn49-Ser162 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | IL15, IL-15MGC9721, interleukin 15, interleukin-15 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction