missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-1RA / IL1RN Protein
A DNA sequence encoding the Homo sapiens (Human) IL-1RA / IL1RN, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8556-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000568.1 | |
IL-1RA / IL1RN Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3557 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
IL1RN | |
Recombinant Protein | |
E. coli | |
RPSGRKSSKM QAFRIWDVNQ KTFYLRNNQL VAGYLQGPNV NLEEKIDVVP IEPHALFLGI HGGKMCLSCV KSGDETRLQL EAVNITDLSE NRKQDKRFAF IRSDSGPTTS FESAACPGWF LCTAMEADQP VSLTNMPDEG VMVTKFYFQE DE |