missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-21 (HEK293 Expressed) Protein

Product Code. 18786255
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
Unit Size:
100µg
10µg
1mg
250µg
50µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18786255 100 μg 100µg
18746315 10 μg 10µg
18755845 250 μg 250µg
18716195 50 μg 50µg
18776185 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18786255 Supplier R&D Systems™ Supplier No. 11393IL100
  • ONLINE EXCLUSIVE PRICE
    1877.40 €
    1695.75 € / 100µg
    Save 181.65 €
    9% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured in a cell proliferation assay using B9 mouse hybridoma cells. The ED50 for this effect is 3.00-30.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HBE4.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 59067
Molecular Weight (g/mol) MolecularWeight-observed: 15-17 kDa, under reducing conditions., MolecularWeight-theroretical: 15 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived human IL-21 protein Gln32-Ser162
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CVID11, IL21, IL-21 Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11
Conjugate Unconjugated
Reconstitution Reconstitute 10 μg size at 100 μg/mL and other sizes (at 500 μg/mL in PBS.)
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-21
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.