missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-22 (HEK293-expressed) Protein

Product Code. 18778806
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
Unit Size:
10µg
1mg
250µg
50µg
This item is not returnable. View return policy

Product Code. 18778806

Brand: R&D Systems™ 11311IL250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured by its ability to induce IL-10 secretion in COLO 205 human colorectal adenocarcinoma cells. Nagalakshmi, M.L. et al. (2004) International Immunopharmacology 4:679. The ED50 for this effect is 0.160-1.60 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9GZX6.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 50616
Molecular Weight (g/mol) M.W. Observed: 28-32 kDa, under reducing conditions., M.W. Theoretical: 17 kDa
Quantity 250 μg
Source Human embryonic kidney cell, HEK293-derived human IL-22 protein Ala34-Ile179
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-22
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.