missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL 23 Protein

A cDNA sequence encoding the IL 23 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10704-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934918

  • 131.00 € / 2µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

51561
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
The activity, as determined by the dose dependent secretion of IL-17 in response to IL-23 using murine splenocytes, the ED50 is 1.9ng/ml.
Human
Untagged
IL-23 was lyophilized from a 0.1microm filtered solution (1.05 mg/ml) containing 1xPBS.
IL 23 Protein
Research Use Only
IL23A
Recombinant Protein
Human (HEK293)
MLGSRAVMLL LLLPWTAQGR AVPGGSSPAW TQCQQLSQKL CTLAWSAHPL VGHMDLREEG DEETTNDVPH IQCGDGCDPQ GLRDNSQFCL QRIHQGLIFY EKLLGSDIFT GEPSLLPDSP VGQLHASLLG LSQLLQPEGH HWETQQIPSL SPSQPWQRLL LRFKILRSLQ AFVAVAARVF AHGAATLSP
Greater than 95.0% as determined by analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers