missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 32A Protein
A cDNA sequence encoding the IL 32A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10714-1mg
Specifications
9235 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human IL-32 alpha activity is measured via the dose-dependent induction of TNF-alpha in the human THP-1 monocytic cell line. | |
Human | |
Untagged | |
IL-32 was lyophilized from a concentrated (1 mg/ml) solution in water containing 50mM sodium Phosphate buffer pH=7.5. |
IL 32A Protein | |
Research Use Only | |
IL32 | |
Recombinant Protein | |
E. coli | |
MCFPKVLSDD MKKLKARMHQ AIERFYDKMQ NAESGRGQVM SSLAELEDDF KEGYLETVAA YYEEQHPELT PLLEKERDGL RCRGNRSPVP DVEDPATEEP GESFCDKSYG APRGDKEELT PQKCSEPQSS K | |
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |