missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-35 Fc Chimera Protein

Product Code. 18785547
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18785547 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18785547 Supplier R&D Systems™ Supplier No. 8608IL050
  • ONLINE EXCLUSIVE PRICE
    567.83 €
    509.25 € / 50µg
    Save 58.58 €
    10% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-35 Fc Chimera Protein is derived from HEK293. The Recombinant Human IL-35 Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 10148
Molecular Weight (g/mol) Observed M.W.: 81-97kDa, reducing conditions, Theoretical M.W.: 73kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human IL-35 protein Human IL-27 beta/EBI3 (Arg21-Lys229) Accession &Num; Q14213 (GGGS)4 Human IL-12 alpha/p35 Arg23-Ser219 Accession &Num; P29459 IEGRMD Human IgG1 Pro100-Lys330)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name IL-35
Gene Symbol EBI3
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human IL-12 R beta 2 Fc Chimera is immobilized at 5μg/mL (100μL/well), the concentration of Recombinant Human IL-35 Fc Chimera that produces 50% optimal binding response is approximately 20-120ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human IL-27 beta/EBI3 (Arg21-Lys229) Accession # Q14213, N-terminus; (GGGS)4; Human IL-12 alpha/p35 Arg23-Ser219 Accession # P29459; IEGRMD; Human IgG1 Pro100-Lys330), C-terminus
Product Line R&D Systems
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.