missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-4 (CHO-expressed) Protein

Product Code. p-7182941
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
This item is not returnable. View return policy

Product Code. 18776416

Brand: R&D Systems™ 6507IL025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. ED50 for this effect is 0.05 to 0.2 ng/mL.

Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.05-0.2 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05112
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3565
Molecular Weight (g/mol) M.W. (Observed): 17-25 kDa, reducing conditions; M.W. (theoretical): 15.0 kDa
Name B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Quantity 25 μg
Source Chinese Hamster Ovary cell line, CHO-derived human IL-4 protein His25-Ser153
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Gene Symbol IL4
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein IL-4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.