missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-7 Protein

Product Code. p-7145914
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18766876

Brand: R&D Systems™ 207IL010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using PHA-activated human peripheral blood lymphocytes (PBL). Yokota, T. et al. (1986) Proc. Natl. Acad. Sci. USA 83:5894. The ED50 for this effect is 0.1-0.5 ng/mL. The specific activity of Recombinant Human IL-7 is approximately 4.4 x 105 IU/μg, which is calibrated against human IL-7 WHO International Standard (NIBSC code: 90/530). Specific activity is for reference purposes only and is not routinely tested.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13232
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3574
Molecular Weight (g/mol) 17 kDa
Quantity 10 μg
Source <EM>E. coli</EM>-derived human IL-7 protein Asp26-His177, with an N-terminal Met
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <0.01 EU per 1 μg of the protein by the LAL method.
Gene Symbol IL7
Conjugate Unconjugated
Cross Reactivity IL-7
Species Human
Reconstitution Reconstitute 5 μg vials at 50 μg/mL in sterile PBS. Reconstitute 10 μg or larger vials (at 100 μg/mL in sterile PBS.)
Buffer Lyophilized from a 0.2μm filtered solution in PBS.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.