missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL12 Protein

A cDNA sequence encoding the IL12 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10722-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15974968

  • 131.00 € / 2µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IL12 Protein
Research Use Only
The activity is determined by the IFN-g ELISA in a using NK-92 cell. The ED50 for this effect is less than or equal to 0.03 ng/ml.
Human
His
IL12 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol.
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Baculovirus
IL12A(p35):RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNASHHH HHH IL12B(p40):IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers