missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL1RL1 Protein
A cDNA sequence encoding the IL1RL1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Gene ID (Entrez) | 9173 |
---|---|
Name | IL1RL1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | IL1RL1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994978
|
enQuireBio™
QP10726-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984978
|
enQuireBio™
QP10726-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974978
|
enQuireBio™
QP10726-0.1MG |
100 μg |
1740.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
9173 | |
Research Use Only | |
IL1RL1 | |
Recombinant Protein | |
Baculovirus | |
KFSKQSWGLE NEALIVRCPR QGKPSYTVDW YYSQTNKSIP TQERNRVFAS GQLLKFLPAA VADSGIYTCI VRSPTFNRTG YANVTIYKKQ SDCNVPDYLM YSTVSGSEKN SKIYCPTIDL YNWTAPLEWF KNCQALQGSR YRAHKSFLVI DNVMTEDAGD YTCKFIHNEN GANYSVTATR SFTVKDEQGF SLFPVIGAPA QNEIKEVEIG KNANLTCSAC FGKGTQFLAA VLWQLNGTKI TDFGEPRIQQ EEGQNQSFSN GLACLDMVLR IADVKEEDLL LQYDCLALNL HGLRRHTVRL SRKNPIDHHS LEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
IL1RL1 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its ability to inhibit proliferation using D10. G4.1 mouse helper T cells. The ED50 for this effect is less or equal to 10ng/ml with IL-33. | |
Human | |
His | |
IL1RL1 protein solution (0.5 mg/ml) contains 10% glycerol and Phosphate Buffered Saline (pH 7.4). |