missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL2 / Interleukin-2 Protein
Recombinant Protein
Brand: enQuireBio™ QP5245-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000577.2 | |
IL2 / Interleukin-2 Protein | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity in the CTLL-2 cell proliferation assay. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
3558 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
E. coli | |
MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFCQSII STLT |