missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL36G / IL1F9 Protein

A DNA sequence encoding the Homo sapiens (Human) IL36G / IL1F9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8125-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042801

  • 505.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001265497.1
IL36G / IL1F9 Protein
Research Use Only
IL36G
Recombinant Protein
E. coli
MRGTPGDADG GGRAVYQSMC KPITGTINDL NQQVWTLQGQ NLVAVPRSDS VTPVTVAVIT CKYPEALEQG RGDPIYLGIQ NPEMCLYCEK VGEQPTLQLK EQKIMDLYGQ PEPVKPFLFY RAKTGRTSTL ESVAFPDWFI ASSKRDQPII LTSELGKSYN TAFELNIND
Greater than 80% as determined by SDS-PAGE.
56300
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers