missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL7R Protein
A cDNA sequence encoding the IL7R was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Name | IL7R Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980389
|
enQuireBio™
QP12429-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15900399
|
enQuireBio™
QP12429-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15990389
|
enQuireBio™
QP12429-50UG |
50 μg |
1740.00 €
50µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL7R Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IL7R protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADLESGYAQN GDLEDAELDD YSFSCYSQLE VNGSQHSLTC AFEDPDVNIT NLEFEICGAL VEVKCLNFRK LQEIYFIETK KFLLIGKSNI CVKVGEKSLT CKKIDLTTIV KPEAPFDLSV VYREGANDFV VTFNTSHLQK KYVKVLMHDV AYRQEKDENK WTHVNLSSTK LTLLQRKLQP AAMYEIKVRS IPDHYFKGFW SEWSPSYYFR TPEINNSSGE MDLEPKSCDK THTCPPCPAP ELLGGPSVFL FPPKPKDTLM ISRTPEVTCV VVDVSHEDPE VKFNWYVDGV EVHNAKTKPR EEQYNSTYRV VSVLTVLHQD WLNGKEYKCK VSNKALPAPI EKTISKAKGQ PREPQVYTLP PSRDELTKNQ VSLTCLVKGF YPSDIAVEWE SNGQPENNYK TTPPVLDSDG SFFLYSKLTV DKSRWQQGNV FSCSVMHEAL HNHYTQKSLS LSPGKHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |