missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ILDR2 Protein

Product Code. 16030621
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16030621 100 μg 100µg
16040621 10 μg 10µg
16080621 50 μg 50µg
16060621 200 μg 200µg
16070621 500 μg 500µg
16050621 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16030621 Supplier enQuireBio™ Supplier No. QP7756ye100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) ILDR2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_955383.1
Gene ID (Entrez) 387597
Name ILDR2 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ILDR2
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence MDRVLLRWIS LFWLTAMVEG LQVTVPDKKK VAMLFQPTVL RCHFSTSSHQ PAVVQWKFKS YCQDRMGESL GMSSTRAQSL SKRNLEWDPY LDCLDSRRTV RVVASKQGST VTLGDFYRGR EITIVHDADL QIGKLMWGDS GLYYCIITTP DDLEGKNEDS VELLVLGRTG LLADLLPSFA VEIMPE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.