missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IMPA1 Protein
A cDNA sequence encoding the IMPA1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12432-5ug
Additional Details : Weight : 0.01000kg
Specifications
IMPA1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MADPWQECMD YAVTLARQAG EVVCEAIKNE MNVMLKSSPV DLVTATDQKV EKMLISSIKE KYPSHSFIGE ESVAAGEKSI LTDNPTWIID PIDGTTNFVH RFPFVAVSIG FAVNKKIEFG VVYSCVEGKM YTARKGKGAF CNGQKLQVSQ QEDITKSLLV TELGSSRTPE TVRMVLSNME KLFCIPVHGI RSVGTAAVNM CLVATGGADA YYEMGIHCWD VAGAGIIVTE AGGVLMDVTG GPFDLMSRRV IAANNRILAE RIAKEIQVIP LQRDDED | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The IMPA1 solution (1 mg/ml) contains 20mM Tris-HCl Buffer (pH 8.0) and 10% Glycerol. |