missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Inhibin alpha A Chain Protein
A cDNA sequence encoding the Inhibin alpha A Chain was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12449-2ug
Additional Details : Weight : 0.01000kg
Specifications
Inhibin alpha A Chain Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Inhibin-A alpha chain is supplied in 20mM Tris HCl (pH 8), 5mM EDTA and 50% glycerol. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |