missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Insulin Protein

A cDNA sequence encoding the Insulin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10732-25mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15994998

  • 179.00 € / 25mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3630
25 mg
< 1.0 EU per ug protein as determined by the LAL method.
27.5 units/mg. Insulin has been evaluated in cell culture (Human Foreskin Fibroblasts). The effective concentration range for use in defined media is 1-5ug/ml. However, the optimum concentration range for a particular application must be determined by the investigator.
Human
Untagged
The protein was lyophilized from 50mM Sodium Chloride solution.
Insulin Protein
Research Use Only
INS
Recombinant Protein
S. cerevisiae
MALWMRLLPL LALLALWGPD PAAAFVNQHL CGSHLVEALY LVCGERGFFY TPKTRREAED LQVGQVELGG GPGAGSLQPL ALEGSLQKRG IVEQCCTSIC SLYQLENYCN
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers